siRNA Details
| Gene Symbol: Slc38a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138832.1 | NP_620187.1 | 14 | 1480 | ||
| XM_006242277.2 | XP_006242339.1 | 14 | 1539 | |||
| XM_006242278.2 | XP_006242340.1 | 15 | 1748 | |||
| XM_006242279.2 | XP_006242341.1 | 15 | 1718 | |||
| XM_006242280.2 | XP_006242342.1 | 14 | 1587 | |||
| XM_006242281.2 | XP_006242343.1 | 14 | 1628 | |||
| GenBank mRNA | AF075704.1 | - | 14 | 1480 | ||
| BC097283.1 | - | 14 | 1400 | 7444889 | ||

Slc38a1
Genomic Map