siRNA Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130750.1 | NP_570106.1 | 12, 13 | 1302 | ||
| GenBank mRNA | AF461260.1 | - | 12, 13 | 1302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asz1| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130750.1 | NP_570106.1 | 12, 13 | 1302 | ||
| GenBank mRNA | AF461260.1 | - | 12, 13 | 1302 | ||