siRNA Details
| Gene Symbol: Fgf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130753.1 | NP_570109.1 | 3 | 397 | ||
| GenBank mRNA | AB078900.1 | - | 3 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf19| Gene Symbol: Fgf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130753.1 | NP_570109.1 | 3 | 397 | ||
| GenBank mRNA | AB078900.1 | - | 3 | 397 | ||