siRNA Details
| Gene Symbol: Acot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130756.3 | NP_570112.3 | 2 | 533 | ||
| GenBank mRNA | AF452100.1 | - | 2 | 183 | ||
| BC078751.1 | - | 2 | 527 | 7114090 | ||
| CV115391.1 | - | 2 | 533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acot8| Gene Symbol: Acot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130756.3 | NP_570112.3 | 2 | 533 | ||
| GenBank mRNA | AF452100.1 | - | 2 | 183 | ||
| BC078751.1 | - | 2 | 527 | 7114090 | ||
| CV115391.1 | - | 2 | 533 | |||