siRNA Details
| Gene Symbol: Fgf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130816.1 | NP_570829.1 | 1 | 105 | ||
| XM_008767766.1 | XP_008765988.1 | 1 | 218 | |||
| GenBank mRNA | AB079263.1 | - | 1 | 105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf11| Gene Symbol: Fgf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130816.1 | NP_570829.1 | 1 | 105 | ||
| XM_008767766.1 | XP_008765988.1 | 1 | 218 | |||
| GenBank mRNA | AB079263.1 | - | 1 | 105 | ||