siRNA Details
| Gene Symbol: Pnma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130820.2 | NP_570833.1 | 1 | 586 | ||
| GenBank mRNA | AF335505.1 | - | 1 | 602 | ||
| BC097291.1 | - | 1 | 562 | 7453131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnma1| Gene Symbol: Pnma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130820.2 | NP_570833.1 | 1 | 586 | ||
| GenBank mRNA | AF335505.1 | - | 1 | 602 | ||
| BC097291.1 | - | 1 | 562 | 7453131 | ||