siRNA Details
| Gene Symbol: Atp6v0c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130823.3 | NP_570836.1 | 2 | 310 | ||
| XM_006245898.1 | XP_006245960.1 | 3 | 811 | |||
| GenBank mRNA | BC063154.1 | - | 2 | 314 | 6918203 | |
| D10874.1 | - | 2 | 251 | |||
| FQ211954.1 | - | 2 | 290 | |||
| FQ212145.1 | - | 2 | 315 | |||
| FQ212559.1 | - | 2 | 294 | |||
| FQ212637.1 | - | 2 | 290 | |||
| FQ212733.1 | - | 2 | 294 | |||
| FQ213105.1 | - | 2 | 294 | |||
| FQ213455.1 | - | 2 | 294 | |||
| FQ214388.1 | - | 2 | 294 | |||
| FQ214503.1 | - | 2 | 315 | |||
| FQ216440.1 | - | 2 | 294 | |||
| FQ220211.1 | - | 2 | 294 | |||
| FQ220713.1 | - | 2 | 315 | |||
| FQ220906.1 | - | 2 | 263 | |||
| FQ221569.1 | - | 2 | 305 | |||
| FQ229281.1 | - | 2 | 295 | |||
| FQ229453.1 | - | 2 | 290 | |||
| FQ229871.1 | - | 2 | 295 | |||
| FQ234225.1 | - | 2 | 315 | |||

Atp6v0c
Genomic Map