siRNA Details
| Gene Symbol: Hadha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130826.2 | NP_570839.2 | 19 | 2043 | ||
| GenBank mRNA | BC091697.1 | - | 19 | 2043 | 7301707 | |
| D16478.1 | - | 19 | 2041 | |||
| FQ218997.1 | - | 5 | 557 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hadha| Gene Symbol: Hadha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130826.2 | NP_570839.2 | 19 | 2043 | ||
| GenBank mRNA | BC091697.1 | - | 19 | 2043 | 7301707 | |
| D16478.1 | - | 19 | 2041 | |||
| FQ218997.1 | - | 5 | 557 | |||