siRNA Details
| Gene Symbol: Nphs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130828.2 | NP_570841.2 | 3 | 454 | ||
| GenBank mRNA | AB091379.1 | - | 3 | 393 | ||
| AF309631.1 | - | 2 | 132 | |||
| AF400653.1 | - | 3 | 174 | |||
| AY039651.1 | - | 3 | 454 | |||
| BC098649.1 | - | 3 | 436 | 7127895 | ||

Nphs2
Genomic Map