siRNA Details
| Gene Symbol: Palm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130829.1 | NP_570842.1 | 149 | |||
| GenBank mRNA | AB058889.1 | - | 149 | |||
| BC072525.1 | - | 190 | 7097177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Palm| Gene Symbol: Palm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130829.1 | NP_570842.1 | 149 | |||
| GenBank mRNA | AB058889.1 | - | 149 | |||
| BC072525.1 | - | 190 | 7097177 | |||