siRNA Details
| Gene Symbol: Hrh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131909.1 | NP_571984.1 | 2 | 313 | ||
| GenBank mRNA | AF358860.1 | - | 2 | 313 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hrh4| Gene Symbol: Hrh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131909.1 | NP_571984.1 | 2 | 313 | ||
| GenBank mRNA | AF358860.1 | - | 2 | 313 | ||
Set Membership: |
|