siRNA Details
| Gene Symbol: TCTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022171.2 | NP_071503.1 | 1, 2 | 442 | ||
| GenBank mRNA | AK000824.1 | - | 1, 2 | 259 | ||
| AK311839.1 | - | 1, 2 | 225 | |||
| BC005157.1 | - | 1, 2 | 408 | 2959904 | ||
| CR457411.1 | - | 1, 2 | 221 | |||
| L41143.1 | - | 1, 2 | 442 | |||

TCTA
Genomic Map