siRNA Details
| Gene Symbol: Anp32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131911.2 | NP_571986.1 | 2 | 243 | ||
| XM_008763695.1 | XP_008761917.1 | 3 | 555 | |||
| GenBank mRNA | BC086508.1 | - | 2 | 243 | 7103152 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anp32b| Gene Symbol: Anp32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131911.2 | NP_571986.1 | 2 | 243 | ||
| XM_008763695.1 | XP_008761917.1 | 3 | 555 | |||
| GenBank mRNA | BC086508.1 | - | 2 | 243 | 7103152 | |