siRNA Details
| Gene Symbol: Anp32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131911.2 | NP_571986.1 | 2 | 365 | ||
| XM_008763695.1 | XP_008761917.1 | 3 | 677 | |||
| GenBank mRNA | AB025581.1 | - | 2 | 202 | ||
| BC086508.1 | - | 2 | 365 | 7103152 | ||

Anp32b
Genomic Map