siRNA Details
| Gene Symbol: Grin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133308.2 | NP_579842.2 | 6 | 2493 | ||
| GenBank mRNA | AF440691.2 | - | 6 | 2493 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin3b| Gene Symbol: Grin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133308.2 | NP_579842.2 | 6 | 2493 | ||
| GenBank mRNA | AF440691.2 | - | 6 | 2493 | ||