siRNA Details
| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133310.2 | NP_579844.2 | 3 | 398 | ||
| GenBank mRNA | BC083923.1 | - | 4 | 603 | ||
| CB616023.1 | - | 3 | 398 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Icmt| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133310.2 | NP_579844.2 | 3 | 398 | ||
| GenBank mRNA | BC083923.1 | - | 4 | 603 | ||
| CB616023.1 | - | 3 | 398 | |||