siRNA Details
| Gene Symbol: Snrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138833.1 | NP_620188.1 | 6 | 1620 | ||
| GenBank mRNA | X89383.1 | - | 6 | 1620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snrk| Gene Symbol: Snrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138833.1 | NP_620188.1 | 6 | 1620 | ||
| GenBank mRNA | X89383.1 | - | 6 | 1620 | ||