siRNA Details
| Gene Symbol: Mutyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133316.1 | NP_579850.1 | 13 | 1211 | ||
| XM_006238626.2 | XP_006238688.1 | 13 | 1239 | |||
| XM_006238627.2 | XP_006238689.1 | 12 | 1136 | |||
| XM_008763987.1 | XP_008762209.1 | 11 | 1197 | |||
| GenBank mRNA | AF478683.1 | - | 13 | 1211 | ||
| BC128728.1 | - | 11 | 1363 | |||

Mutyh
Genomic Map