siRNA Details
| Gene Symbol: TCTE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174910.1 | NP_777570.1 | 3 | 518 | ||
| XM_006715554.3 | XP_006715617.1 | 3 | 377 | |||
| XM_011536093.2 | XP_011534395.1 | 3 | 557 | |||
| GenBank mRNA | AF519569.1 | - | 3 | 518 | ||
| BC113639.1 | - | 3 | 459 | 8322691 | ||

TCTE3
Genomic Map