siRNA Details
| Gene Symbol: Khdrbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133318.1 | NP_579852.1 | 4 | 556 | ||
| XM_008766952.1 | XP_008765174.1 | 4 | 576 | |||
| GenBank mRNA | AF305618.1 | - | 4 | 556 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdrbs2| Gene Symbol: Khdrbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133318.1 | NP_579852.1 | 4 | 556 | ||
| XM_008766952.1 | XP_008765174.1 | 4 | 576 | |||
| GenBank mRNA | AF305618.1 | - | 4 | 556 | ||