siRNA Details
| Gene Symbol: Ndel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133320.1 | NP_579854.1 | 4 | 390 | ||
| XM_006246571.2 | XP_006246633.1 | 4 | 799 | |||
| XM_006246572.2 | XP_006246634.1 | 4 | 792 | |||
| XM_006246573.2 | XP_006246635.1 | 4 | 540 | |||
| XM_006246574.2 | XP_006246636.1 | 4 | 425 | |||
| XM_006246575.2 | XP_006246637.1 | 4 | 903 | |||
| XM_006246576.2 | XP_006246638.1 | 4 | 793 | |||
| XM_006246578.2 | XP_006246640.1 | 4 | 539 | |||
| GenBank mRNA | AY008298.1 | - | 4 | 390 | ||
| BC070909.1 | - | 4 | 480 | 7103650 | ||

Ndel1
Genomic Map