siRNA Details
| Gene Symbol: Kcnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172041.2 | NP_742038.2 | 7 | 1274 | ||
| NM_172042.1 | NP_742039.1 | 7 | 1262 | |||
| XM_006250433.2 | XP_006250495.1 | 7 | 1485 | |||
| XM_006250434.2 | XP_006250496.1 | 7 | 1864 | |||
| GenBank mRNA | AF325671.1 | - | 7 | 1262 | ||
| AF385402.1 | - | 7 | 1369 | |||
| AY555072.1 | - | 7 | 1225 | |||
| AY555073.1 | - | 6 | 1059 | |||
| AY695826.1 | - | 7 | 1219 | |||
| AY727922.1 | - | 7 | 1217 | |||
| DQ403851.1 | - | 7 | 1274 | |||

Kcnk2
Genomic Map