siRNA Details
| Gene Symbol: Zfp347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133390.1 | NP_596881.1 | 5 | 3222 | ||
| GenBank mRNA | AB047637.1 | - | 5 | 3222 | ||
| AB047638.1 | - | 2610 | ||||
| Gene Symbol: LOC102551257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252988.2 | XP_006253050.1 | 4 | 2108 | ||
| XM_006252989.2 | XP_006253051.1 | 4 | 1994 | |||

Zfp347
Genomic Map