siRNA Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133392.1 | NP_596883.1 | 7 | 856 | ||
| XM_006244908.2 | XP_006244970.1 | 7 | 1625 | |||
| GenBank mRNA | AB070349.1 | - | 7 | 856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk17b| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133392.1 | NP_596883.1 | 7 | 856 | ||
| XM_006244908.2 | XP_006244970.1 | 7 | 1625 | |||
| GenBank mRNA | AB070349.1 | - | 7 | 856 | ||