siRNA Details
Gene Symbol: Serinc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ103710.1 | - | 11 | 1197 | ||
L20319.1 | - | 11 | 1318 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Serinc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ103710.1 | - | 11 | 1197 | ||
L20319.1 | - | 11 | 1318 |