siRNA Details
| Gene Symbol: Tesk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133396.2 | NP_596887.1 | 5, 6 | 901 | ||
| XM_006238628.2 | XP_006238690.1 | 5, 6 | 1132 | |||
| GenBank mRNA | AB049402.1 | - | 5, 6 | 902 | ||
| BC081737.1 | - | 6, 7 | 1152 | |||
| BC128727.1 | - | 5, 6 | 907 | 7111312 | ||

Tesk2
Genomic Map