siRNA Details
| Gene Symbol: Erg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133397.2 | NP_596888.1 | 9 | 1212 | ||
| XM_006248142.2 | XP_006248204.1 | 10 | 1443 | |||
| XM_006248143.2 | XP_006248205.1 | 9 | 1374 | |||
| XM_006248145.2 | XP_006248207.1 | 8 | 1301 | |||
| GenBank mRNA | AB031088.1 | - | 9 | 1212 | ||
| BC072519.1 | - | 9 | 1192 | 7105647 | ||
| FQ217228.1 | - | 9 | 1231 | |||

Erg
Genomic Map