siRNA Details
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133399.2 | NP_596890.2 | 16 | 2452 | ||
| GenBank mRNA | AF395897.1 | - | 4 | 340 | ||
| CR754046.1 | - | 3 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3ca| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133399.2 | NP_596890.2 | 16 | 2452 | ||
| GenBank mRNA | AF395897.1 | - | 4 | 340 | ||
| CR754046.1 | - | 3 | 268 | |||