siRNA Details
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133399.2 | NP_596890.2 | 3, 4 | 810 | ||
| GenBank mRNA | CK366305.1 | - | 1, 2 | 42 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3ca| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133399.2 | NP_596890.2 | 3, 4 | 810 | ||
| GenBank mRNA | CK366305.1 | - | 1, 2 | 42 | ||