siRNA Details
| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133400.1 | NP_596891.1 | 7 | 902 | ||
| XM_006231272.2 | XP_006231334.1 | 7 | 1073 | |||
| GenBank mRNA | AF290984.1 | - | 7 | 902 | ||
| AY028945.1 | - | 6 | 800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A1cf| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133400.1 | NP_596891.1 | 7 | 902 | ||
| XM_006231272.2 | XP_006231334.1 | 7 | 1073 | |||
| GenBank mRNA | AF290984.1 | - | 7 | 902 | ||
| AY028945.1 | - | 6 | 800 | |||