siRNA Details
| Gene Symbol: Lonp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133404.1 | NP_596895.1 | 16 | 2398 | ||
| GenBank mRNA | AB064323.1 | - | 16 | 2398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lonp1| Gene Symbol: Lonp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133404.1 | NP_596895.1 | 16 | 2398 | ||
| GenBank mRNA | AB064323.1 | - | 16 | 2398 | ||