siRNA Details
| Gene Symbol: Cry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133405.2 | NP_596896.2 | 3, 4 | 458 | ||
| GenBank mRNA | AY033877.1 | - | 3, 4 | 468 | ||
| BC166416.1 | - | 3, 4 | 458 | 9033484 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cry2| Gene Symbol: Cry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133405.2 | NP_596896.2 | 3, 4 | 458 | ||
| GenBank mRNA | AY033877.1 | - | 3, 4 | 468 | ||
| BC166416.1 | - | 3, 4 | 458 | 9033484 | ||