siRNA Details
| Gene Symbol: Agpat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133406.1 | NP_596897.1 | 2 | 344 | ||
| XM_006227856.2 | XP_006227918.1 | 2 | 1059 | |||
| XM_006227857.2 | XP_006227919.1 | 2 | 713 | |||
| GenBank mRNA | AB067572.1 | - | 2 | 344 | ||
| BC086992.1 | - | 2 | 263 | 7191675 | ||

Agpat4
Genomic Map