siRNA Details
| Gene Symbol: DYNLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291602.1 | NP_001278531.1 | 2, 3 | 222 | ||
| NM_006519.3 | NP_006510.1 | 3, 4 | 264 | |||
| GenBank mRNA | AK315601.1 | - | 3, 4 | 221 | ||
| BC029412.1 | - | 3, 4 | 217 | 4074224 | ||
| BC105588.1 | - | 3, 4 | 213 | 6723966 | ||
| CR456931.1 | - | 3, 4 | 186 | |||
| D50663.1 | - | 3, 4 | 198 | |||
| EU862237.1 | - | 3, 4 | 190 | |||
| HY017024.1 | - | 2, 3 | 175 | |||
| HY171700.1 | - | 3, 4 | 267 | |||
| U56255.1 | - | 3, 4 | 186 | |||

DYNLT1
Genomic Map