siRNA Details
| Gene Symbol: Ilk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133409.2 | NP_596900.1 | 9, 10 | 1039 | ||
| GenBank mRNA | AF329194.1 | - | 9, 10 | 976 | ||
| BC062406.1 | - | 9, 10 | 1039 | 5623251 | ||
| FQ233927.1 | - | 3, 4 | 366 | |||

Ilk
Genomic Map