siRNA Details
| Gene Symbol: Abcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133411.1 | NP_596902.1 | 19, 20 | 2463 | ||
| XM_008770939.1 | XP_008769161.1 | 19, 20 | 2851 | |||
| GenBank mRNA | AF534126.1 | - | 19, 20 | 2474 | ||
| AY533524.1 | - | 19, 20 | 2463 | |||
| AY533525.1 | - | 18, 19 | 2352 | |||

Abcc4
Genomic Map