siRNA Details
| Gene Symbol: Slco6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133412.1 | NP_596903.1 | 12 | 2089 | ||
| GenBank mRNA | AF321415.1 | - | 12 | 2089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco6b1| Gene Symbol: Slco6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133412.1 | NP_596903.1 | 12 | 2089 | ||
| GenBank mRNA | AF321415.1 | - | 12 | 2089 | ||