siRNA Details
Gene Symbol: Clcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133414.1 | NP_596905.1 | 6 | 739 | ||
XM_006233127.1 | XP_006233189.1 | 6 | 778 | |||
XM_006233128.2 | XP_006233190.1 | 6 | 837 | |||
XM_008761365.1 | XP_008759587.1 | 6 | 837 | |||
XM_008761366.1 | XP_008759588.1 | 5 | 676 | |||
GenBank mRNA | AB052919.1 | - | 4 | 595 | ||
AB052920.1 | - | 4 | 595 | |||
AB052921.1 | - | 5 | 697 | |||
AB052922.1 | - | 6 | 739 | |||
AB052923.1 | - | 6 | 759 | |||
AF117330.1 | - | 6 | 727 | |||
BC081736.1 | - | 6 | 815 | 7128078 |
