siRNA Details
Gene Symbol: Necab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133415.1 | NP_596906.1 | 11 | 962 | ||
XM_006255703.2 | XP_006255765.1 | 9 | 2028 | |||
GenBank mRNA | AF193757.1 | - | 11 | 962 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Necab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133415.1 | NP_596906.1 | 11 | 962 | ||
XM_006255703.2 | XP_006255765.1 | 9 | 2028 | |||
GenBank mRNA | AF193757.1 | - | 11 | 962 |