siRNA Details
| Gene Symbol: Hrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133428.2 | NP_596919.1 | 4 | 452 | ||
| GenBank mRNA | AB055895.1 | - | 4 | 452 | ||
| BC089779.1 | - | 4 | 441 | 7371893 | ||
| FQ210661.1 | - | 4 | 452 | |||
| FQ219046.1 | - | 2 | 56 | |||

Hrg
Genomic Map