siRNA Details
| Gene Symbol: Atp12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301664.1 | NP_001288593.1 | 16 | 2871 | ||
| NM_133517.2 | NP_598201.2 | 17 | 2730 | |||
| GenBank mRNA | M90398.1 | - | 17 | 2654 | ||
| U94912.1 | - | 17 | 2726 | |||
| U94913.1 | - | 16 | 2870 | |||

Atp12a
Genomic Map