siRNA Details
| Gene Symbol: Dnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133525.1 | NP_598209.1 | 3 | 315 | ||
| GenBank mRNA | U82591.1 | - | 3 | 315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnph1| Gene Symbol: Dnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133525.1 | NP_598209.1 | 3 | 315 | ||
| GenBank mRNA | U82591.1 | - | 3 | 315 | ||