siRNA Details
| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133527.1 | NP_598211.1 | 4 | 472 | ||
| XM_006229842.2 | XP_006229904.1 | 4 | 593 | |||
| GenBank mRNA | AF219904.1 | - | 4 | 472 | ||
| BC157811.1 | - | 3 | 455 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Folr1| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133527.1 | NP_598211.1 | 4 | 472 | ||
| XM_006229842.2 | XP_006229904.1 | 4 | 593 | |||
| GenBank mRNA | AF219904.1 | - | 4 | 472 | ||
| BC157811.1 | - | 3 | 455 | |||