siRNA Details
| Gene Symbol: Mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133528.1 | NP_598212.1 | 7 | 539 | ||
| GenBank mRNA | AJ132008.1 | - | 7 | 539 | ||
| BC085708.1 | - | 7 | 560 | 7193103 | ||
| FQ229796.1 | - | 7 | 555 | |||
| FQ233766.1 | - | 7 | 562 | |||

Mob4
Genomic Map