siRNA Details
| Gene Symbol: Mmp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133530.1 | NP_598214.1 | 8, 9 | 1229 | ||
| GenBank mRNA | M60616.1 | - | 8, 9 | 1202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp13| Gene Symbol: Mmp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133530.1 | NP_598214.1 | 8, 9 | 1229 | ||
| GenBank mRNA | M60616.1 | - | 8, 9 | 1202 | ||