siRNA Details
Gene Symbol: Ccr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133532.2 | NP_598216.2 | 1 | 395 | ||
XM_006243915.2 | XP_006243977.1 | 2 | 582 | |||
GenBank mRNA | AF432872.2 | - | 1 | 395 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133532.2 | NP_598216.2 | 1 | 395 | ||
XM_006243915.2 | XP_006243977.1 | 2 | 582 | |||
GenBank mRNA | AF432872.2 | - | 1 | 395 |