siRNA Details
| Gene Symbol: Ccr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133532.2 | NP_598216.2 | 1 | 519 | ||
| XM_006243915.2 | XP_006243977.1 | 2 | 706 | |||
| GenBank mRNA | AF432872.2 | - | 1 | 519 | ||
| AF432873.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr4| Gene Symbol: Ccr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133532.2 | NP_598216.2 | 1 | 519 | ||
| XM_006243915.2 | XP_006243977.1 | 2 | 706 | |||
| GenBank mRNA | AF432872.2 | - | 1 | 519 | ||
| AF432873.1 | - | 1 | 114 | |||