siRNA Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133533.2 | NP_598217.1 | 4 | 541 | ||
| GenBank mRNA | BC088115.1 | - | 4 | 541 | 7325406 | |
| FQ226266.1 | - | 4 | 538 | |||
| FQ227061.1 | - | 4 | 536 | |||
| FQ230261.1 | - | 4 | 532 | |||
| FQ232054.1 | - | 4 | 532 | |||
| FQ232680.1 | - | 4 | 532 | |||
| FQ232790.1 | - | 4 | 555 | |||
| FQ233507.1 | - | 4 | 530 | |||
| FQ234082.1 | - | 4 | 515 | |||
| FQ234487.1 | - | 4 | 530 | |||

Cd79b
Genomic Map