siRNA Details
| Gene Symbol: Rab3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133536.2 | NP_598220.1 | 2 | 284 | ||
| XM_008760719.1 | XP_008758941.1 | 2 | 449 | |||
| GenBank mRNA | D78197.1 | - | 2 | 261 | ||
| U54807.1 | - | 2 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab3c| Gene Symbol: Rab3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133536.2 | NP_598220.1 | 2 | 284 | ||
| XM_008760719.1 | XP_008758941.1 | 2 | 449 | |||
| GenBank mRNA | D78197.1 | - | 2 | 261 | ||
| U54807.1 | - | 2 | 288 | |||