siRNA Details
| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133537.2 | NP_598221.2 | 3 | 344 | ||
| GenBank mRNA | X13309.1 | - | 3 | 333 | ||
| Gene Symbol: Wfdc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727786.2 | XP_002727832.1 | 3 | 339 | ||

Wfdc18
Genomic Map